missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFRA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | GFRA2 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891515
|
Proteintech
21973-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881515
|
Proteintech
21973-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes all the three isoforms.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.Specifications
GFRA2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O00451, O08842 | |
14586, 2675 | |
GFRA2 Fusion Protein Ag16893 | |
Primary | |
-20°C | |
GFRA2 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GDNF family receptor alpha 2, GDNF receptor alpha 2, GDNF receptor beta, GDNFR alpha 2, GDNFR beta, GDNFRB, GFR alpha 2, GFRA2, Neurturin receptor alpha, NRTNR ALPHA, NTNR alpha, NTNRA, RET ligand 2, RETL2, TRNR2 | |
Gfra2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |