missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 907 Germination and Seed Test Papers, Sheet

For germination assays of seed specimens according to various methods such as Neeb and Jacobsen.

Brand:  Fisherbrand™ 0907F00002

 View more versions of this product

Product Code. 11799905

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Wetstrength paper 160g/m²
  • Absorbent
  • Free from chlorine
  • pH must be between 6.0 and 7.5
  • For small seeds such as grasses and flowers
  • White in color
Specifications

Specifications

160 g/m2
Paper
100
0.38 mm
6.10 in.
White
Germination assays of seed specimen
6 to 7.5
Germination and Seed Test Paper
155 x 175mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers