Learn More
Vilber Lourmat™ Germicidal UV Lamp
Brand: Vilber Lourmat™ 312134011
This item is not returnable.
View return policy
Specifications
Germicidal UV Lamp | |
120 W | |
140 mm | |
110 mm | |
1290 mm | |
1 Ea. |
230 V | |
254 nm | |
120 W, 230 V, 50 Hz | |
50 Hz | |
219 mW/cm2 |