Learn More
Vilber Lourmat™ Germicidal UV Lamp
Brand:  Vilber Lourmat™ 3121 3151 1
 View more versions of this product
Product Code. 11766728
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Germicidal UV Lamp
45 W
140 mm
110 mm
500 mm
1 Ea.
230 V
254 nm
45 W, 230 V, 50 Hz
50 Hz
95 mW/cm2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products