missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm, Thermo Scientific Chemicals
Form: Electrical Resistivity: 40 ohm-cm
352.00 € - 1252.00 €
Chemical Identifiers
CAS | 7440-56-4 |
---|---|
Molecular Formula | Ge |
Molecular Weight (g/mol) | 72.63 |
MDL Number | MFCD00085310 |
InChI Key | GNPVGFCGXDBREM-UHFFFAOYSA-N |
Synonym | element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder |
PubChem CID | 6326954 |
ChEBI | CHEBI:30441 |
IUPAC Name | germanium |
SMILES | [Ge] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381248
|
Thermo Scientific Alfa Aesar
010192.TK |
24 g |
352.00 €
24g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391248
|
Thermo Scientific Alfa Aesar
010192.79 |
120 g |
1252.00 €
120g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-56-4 | |
72.63 | |
GNPVGFCGXDBREM-UHFFFAOYSA-N | |
6326954 | |
germanium |
Ge | |
MFCD00085310 | |
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder | |
CHEBI:30441 | |
[Ge] |
Specifications
7440-56-4 | |
Ingot | |
Ge | |
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder | |
[Ge] | |
72.63 | |
CHEBI:30441 | |
Silver-colored ingot |
Silver | |
99.9999% (Metals basis, 95% N type minimum) | |
MFCD00085310 | |
GNPVGFCGXDBREM-UHFFFAOYSA-N | |
germanium | |
6326954 | |
Odorless | |
Germanium |
Safety and Handling
missing translation for 'einecsNumber' : 231-164-3
missing translation for 'rtecsNumber' : LY5200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only