missing translation for 'onlineSavingsMsg'
Learn More

Gerber Test Sulphuric Acid FMT d. 1.815-1.825, Reagecon
BIOPHARMA

Gerber Test Sulphuric Acid FMT d. 1.815-1.825

40.10 € - 211.00 €

Specifications

For Use With (Application) Gerber Test
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11563122
BIOPHARMA
View Documents
Reagecon
SUFMTJ
2.5 L PE-HD Safe Grip Bottle
40.10 €
Each
Please to purchase this item. Need a web account? Register with us today!
11553122
View Documents
Reagecon
SUFMT25
25 L HDPE Blue Drum
211.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gerber Test Sulphuric Acid FMT d. 1.815-1.825
Specifications

Specifications

Gerber Test
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SULPHURIC ACID FMT

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.