missing translation for 'onlineSavingsMsg'
Learn More

GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

132.87 € - 359.47 €

Specifications

Antigen GEFT
Concentration 0.5 mg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883694
View Documents
Proteintech
14839-1-AP-20UL
20 μL
132.87 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16873694
View Documents
Proteintech
14839-1-AP-150UL
150 μL
359.47 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
Specifications

Specifications

GEFT
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
Q6P720, Q86VW2, Q9CWR0
115557, 314904, 52666
Peptide
Primary
-20°C
ARHGEF25
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor
Arhgef25
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.