missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Quartz Microvolume Cuvette

Quartz cell with 40μL working volume and reduced height.

Brand:  Biochrom 80-2076-38

Product Code. 10427294

  • 872.00 € / 50µL
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 10 mm Quartz cuvette
  • 40 μL working volume, reduced height
  • For use with Ultrospec 2100 Pro
  • Requires 80-2106-10 cylindrical cell holder
Specifications

Specifications

40 μL
10 mm
Spectrophotometers, Photometers
Spectrophotometric Measurements
12 mm
Quartz
190 to 1100 nm
12 mm
Square or Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers