Learn More
Cytiva TE 77 PWR Semi-Dry Transfer Unit
The TE 77 PWR offers the convenience of a built-in power supply, and a unique automatic stopping feature. The transfer is stopped when buffer gets depleted, preventing overheating.
Brand:  Cytiva 11-0013-42
Product Code. 10630996
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Tested for use with Amersham ECL Western blotting reagents
  • Built-in power supply
  • Use minimal buffer - only enough to saturate blotting papers and membranes
  • Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers
  • Use low current and voltage for transfers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amersham™ ECL™ Semi Dry Blotter
TE 77 PWR Semi Dry Transfer Unit