missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva His GraviTrap™ Kit
For fast and simple purification of histidine-tagged proteins
Brand: Cytiva 28-4013-51
Includes: Two packs of His GraviTrap and one pack of His buffer kit
Description
- Binding capacity: approximately 40mg histidine-tagged protein/column
- Bed volume: 1mL
- Column hardware material: polypropylene (barrel), polyethylene (frits)
Specifications
1 mL | |
His GraviTrap kit | |
Polypropylene (PP) barrel, Polyethylene (PE) frits | |
10 Columns | |
Gravity Flow Column |
Approx. 40 mg histidine-tagged protein/column | |
2 Packs of His GraviTrap and 1 Pack of His Buffer Kit | |
4°C to 30°C, 20% Ethanol | |
For fast and simple purification of histidine-tagged* proteins |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger||Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, Nickel, imidazole