missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva High Purity Solution ddNTPs
Use in any application that requires ddNTPs of high purity. Cytiva Life Sciences™ High Purity Solution ddNTPs are available either as individual products or in a convenient set.
Specifications
Color | Colorless |
---|---|
Format | Liquid |
For Use With (Application) | Dideoxy sequencing, genotyping reactions |
Purity | ≥98% |
Concentration | 100 mM |
Description
- Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
- High purity: >98% triphosphate
Dideoxy sequencing, Genotyping reactions
Spécification
Colorless | |
Dideoxy sequencing, genotyping reactions | |
100 mM |
Liquid | |
≥98% | |
4 μmol (40 μL) |