missing translation for 'onlineSavingsMsg'
Learn More

Cytiva cGMP Direct BioTrak™ EIA

Use for direct measurement of cGMP in cell lysates. Cytiva Life Sciences™ cGMP Direct BioTrak™ EIA is a novel lysis reagent that allows cGMP measurement without any need for removal of extraction reagents.

Brand:  Cytiva RPN226

Product Code. 10762257

  • 1133.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Dual range: 2–512 fmol/well [acetylation], 50–12 800 fmol/well [non-acetylation]
  • Sensitivity: 14pg/mL [acetylation], 161pg/mL [non-acetylation]
  • One-day protocol
  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents

TRUSTED_SUSTAINABILITY
Specifications

Specifications

14pg/mL
96-well Microplate
Acetylation reagents, Lysis reagents
2°C to 8°C
cGMP direct Biotrak EIA
Enzyme immunoassay for direct cGMP measurement
96 Determinations
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3

SDS
Documents

Documents

Certificates
Special Offers

Special Offers