missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ GC Deactivated Inlet Liner Splitless Straight-Through Liner Quartz Wool For Agilent 7890

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09200401

Product Code. 12927141

  • 51.35 € / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

    Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Agilent Technologies instruments
Specifications

Specifications

78.5 mm
2 mm
Agilent Technologies Instruments
GC Inlet Liner, Split / Splitless Quartz, Straight-Through Inlet Liner
6.1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers