Learn More
Manutan Galvanised wire rope, light duty, 175kg WLL capacity, 50m long coil
Brand:  Manutan A340130
Encompass_Preferred
Product Code. 12808658
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Galvanised wire rope manufactured to current British Standards. Light duty rope, resistant to corrosion, suitable for garage doors and security cables. WLL based on 5:1 safety factor.
  • Diameter: 4mm
  • WLL: 175kg