Learn More
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals
Requires overnight shipping with cold pack
228.14 € - 2253.21 €
Chemical Identifiers
CAS | 7440-55-3 |
---|---|
Molecular Formula | Ga |
Molecular Weight (g/mol) | 69.72 |
MDL Number | MFCD00134045 |
InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
Synonym | gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom |
PubChem CID | 5360835 |
ChEBI | CHEBI:49631 |
IUPAC Name | gallium |
SMILES | [Ga] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385749
|
Thermo Scientific Alfa Aesar
010185.14 |
25 g |
228.14 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301238
|
Thermo Scientific Alfa Aesar
010185.18 |
50 g | N/A | ||||||
11321238
|
Thermo Scientific Alfa Aesar
010185.30 |
250 g | N/A | ||||||
11311238
|
Thermo Scientific Alfa Aesar
010185.36 |
500 g |
2253.21 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
5360835 | |
gallium |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
CHEBI:49631 | |
[Ga] |
Specifications
Silver to Gray | |
Silver Gray ingot | |
Ga | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
Gallium |
7440-55-3 | |
99.9999% (Metals basis) | |
MFCD00134045 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
Ingot |
Safety and Handling
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 231-163-8
- Gallium ingot
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.