Learn More
Gallium ingot, 99.999% (metals basis), Thermo Scientific Chemicals
302.00 € - 2370.00 €
Chemical Identifiers
CAS | 7440-55-3 |
---|---|
Molecular Formula | Ga |
Molecular Weight (g/mol) | 69.72 |
MDL Number | MFCD00134045 |
InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
Synonym | gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom |
PubChem CID | 5360835 |
ChEBI | CHEBI:49631 |
IUPAC Name | gallium |
SMILES | [Ga] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420987
|
Thermo Scientific Alfa Aesar
010187.14 |
25 g |
302.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361238
|
Thermo Scientific Alfa Aesar
010187.18 |
50 g |
431.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430987
|
Thermo Scientific Alfa Aesar
010187.30 |
250 g |
1432.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371238
|
Thermo Scientific Alfa Aesar
010187.36 |
500 g |
2370.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
5360835 | |
gallium |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
CHEBI:49631 | |
[Ga] |
Specifications
Gray-Silver | |
Ga | |
UN2803 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
99.999% | |
(metals basis) |
7440-55-3 | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
Ingot | |
Gallium ingot |
Safety and Handling
GHS H Statement
H290
May be corrosive to metals.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM
missing translation for 'einecsNumber' : 231-163-8
missing translation for 'rtecsNumber' : LW8600000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Requires overnight shipping with cold pack; Store at 2°C to 8°C
Recommended Storage : Keep cold
- Gallium ingot
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
RUO – Research Use Only