missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11304298
Click to view available options
Quantity:
0.2 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Gallium(III) sulfide is used in semiconductor materials, functional ceramic materials, fluorescent powder and photoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-22-5
Molecular Formula Ga2S3
Molecular Weight (g/mol) 235.63
MDL Number MFCD00016106
InChI Key BVSHTEBQPBBCFT-UHFFFAOYSA-N
IUPAC Name digallium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Ga+3].[Ga+3]

Specifications

Melting Point 1255°C
Physical Form Powder
Quantity 0.2 g
UN Number UN3260
Solubility Information It dissolves in water slowly,which is easy to dissolve in concentrated alkaline to get gallate salts. It is also dissolves in HCL and nitric acid.
Formula Weight 235.63
Percent Purity 99.999%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.77 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'