Learn More
Galena, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Description
Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Black |
Physical Form | Powder, Flake, Crystalline, Beads, Lump, Etc. |
Quantity | 25 g |
Solubility Information | Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide. |
Formula Weight | 239.25 |
Odor | Odorless |
Chemical Name or Material | Galena, naturally occurring mineral, grains, approximately 0.06-0.19 in. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.