missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14979-1-AP-20UL
This item is not returnable.
View return policy
Description
The monoclonal antibody B2C10 reacts with galectin-3, a 30 kDa protein. Galectin-3 is a member of the galectin family. The protein is compose of three domains: a small amino-terminal domain, a carboxyl-terminal carbohydrate recognition domain (CRD) and amino-terminal domain containing repeating elements. Galectin-3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. The expression is also affected by neoplastic transformation: up-regulated in certain types of lymphomas and thyroid carcinoma, while down-regulated in other types of maligcies, such as colon, breast, ovarian and uterine carcinomas.Specifications
Galectin-3 | |
Polyclonal | |
Unconjugated | |
LGALS3 | |
35 kDa lectin, CBP 35, CBP35, Gal 3, GAL3, Galactose specific lectin 3, Galactoside binding protein, GALBP, Galectin 3, Galectin-3, GALIG, IgE binding protein, L 31, Laminin binding protein, Lectin L 29, LGALS2, LGALS3, Mac 2 antigen, MAC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16854, 3958, 83781 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08699, P16110, P17931 | |
Lgals3 | |
Galectin-3 Fusion Protein Ag6891 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction