missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals

42.21 € - 150.75 €

Chemical Identifiers

CAS 25764-15-2
Molecular Formula GdN
Molecular Weight (g/mol) 171.26
MDL Number MFCD00049943
InChI Key FLATXDRVRRDFBZ-UHFFFAOYSA-N
Synonym gadolinium nitride, gadolinium iii nitride, gadoliniumylidyneamine
PubChem CID 117632
IUPAC Name azanylidynegadolinium
SMILES N#[Gd]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15462747
SDS View Documents
Thermo Scientific Alfa Aesar
046709.03
1 g
42.21 €
1g
Please to purchase this item. Need a web account? Register with us today!
15472747
SDS View Documents
Thermo Scientific Alfa Aesar
046709.06
5 g
150.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 247-250-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.