Learn More
Gadolinium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
For vacuum deposition
109.55 € - 454.26 €
Chemical Identifiers
CAS | 13765-26-9 |
---|---|
Molecular Formula | F3Gd |
Molecular Weight (g/mol) | 214.25 |
MDL Number | MFCD00016113 |
InChI Key | TYIZUJNEZNBXRS-UHFFFAOYSA-K |
IUPAC Name | gadolinium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[Gd+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304118
|
Thermo Scientific Alfa Aesar
041412.09 |
10 g |
109.55 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314118
|
Thermo Scientific Alfa Aesar
041412.18 |
50 g |
454.26 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13765-26-9 | |
214.25 | |
TYIZUJNEZNBXRS-UHFFFAOYSA-K | |
[F-].[F-].[F-].[Gd+3] |
F3Gd | |
MFCD00016113 | |
gadolinium(3+) trifluoride |
Specifications
1231°C | |
White | |
F3Gd | |
Insoluble in water. | |
[F-].[F-].[F-].[Gd+3] | |
214.25 | |
99.9% | |
Hygroscopic | |
(REO) |
13765-26-9 | |
Pieces | |
MFCD00016113 | |
TYIZUJNEZNBXRS-UHFFFAOYSA-K | |
gadolinium(3+) trifluoride | |
214.25 | |
Odorless | |
7.1 g/mL | |
Gadolinium(III) fluoride, Anhydrous |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 237-369-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.