Learn More
Gadolinium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals
25.30 € - 211.00 €
Chemical Identifiers
CAS | 100587-93-7 |
---|---|
Molecular Formula | C6H9GdO6 |
Molecular Weight (g/mol) | 334.38 |
MDL Number | MFCD00150116 |
InChI Key | LYQGMALGKYWNIU-UHFFFAOYSA-K |
IUPAC Name | gadolinium(3+) triacetate |
SMILES | [Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339399
|
Thermo Scientific Alfa Aesar
089158.06 |
5 g |
25.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349399
|
Thermo Scientific Alfa Aesar
089158.14 |
25 g |
74.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329399
|
Thermo Scientific Alfa Aesar
089158.22 |
100 g |
211.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gadolinium(III) acetate hydrate is used as a laboratory reagent, catalysts, optical glasses, structural ceramics, electrical components and photo-optical material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100587-93-7 | |
334.38 | |
LYQGMALGKYWNIU-UHFFFAOYSA-K | |
[Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H9GdO6 | |
MFCD00150116 | |
gadolinium(3+) triacetate |
Specifications
(decomposition) | |
White | |
99.9% (REO) | |
MFCD00150116 | |
LYQGMALGKYWNIU-UHFFFAOYSA-K | |
gadolinium(3+) triacetate | |
334.39 (Anhydrous) | |
Hygroscopic | |
Gadolinium(III) acetate hydrate |
100587-93-7 | |
Crystals | |
C6H9GdO6 | |
Soluble in water. | |
[Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
334.38 | |
Odorless | |
1.611 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 240-204-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only