missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD65 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21760-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to GAD2.
GAD65 (glutamic acid decarboxylase-65) are present in autoimmune disorders such as insulin-dependent (type1) diabetes mellitus (IDDM), stiff man syndrome and polyendocrine autoimmune disease. Auto-antibodies to GAD65 are present in 60-70% of individuals with newly diagnosed IDDM and thus are important markers for disease activity. These auto-antibodies usually recognize conformation-dependent regions on GAD65 and rarely bind to the second isoform, GAD67. Auto-antibodies to GAD67 are found in only 15% of recent-onset IDDM patients and most of this binding can be blocked with GAD65, suggesting shared epitopes between the two isoforms of GAD.Specifications
GAD65 | |
Polyclonal | |
Unconjugated | |
GAD2 | |
GAD 65, GAD2, GAD65, Glutamate decarboxylase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14417, 24380, 2572 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48320, Q05329, Q05683 | |
Gad2 | |
GAD65 Fusion Protein Ag16251 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction