missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAPL1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E10F6, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | GABARAPL1 |
---|---|
Clone | 1E10F6 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858355
|
Proteintech
66458-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848355
|
Proteintech
66458-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.Specifications
GABARAPL1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1, GEC 1, GEC1 | |
Gabarapl1 | |
IgG1 | |
Protein G | |
Antibody |
1E10F6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat, Porcine, Human | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
100462751, 23710, 57436, 689161 | |
GABARAPL1 Fusion Protein Ag1473 | |
Primary | |
-20°C | |
GABARAPL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title