missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22439-1-AP-20UL
This item is not returnable.
View return policy
Description
Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system. It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. GABARAP (GABAA receptor-associated protein) links GABAA receptors to the cytoskeleton and may play a role in intracellular transport of GABAA receptors and its interaction with the cytoskeleton. GABARAP belongs to the MAP1 or ATG8 like family and recent studies show that MAPK15/ERK8 is acting through interaction with ATG8 family proteins to regulate autophagy.Specifications
GABARAP | |
Polyclonal | |
Unconjugated | |
GABARAP | |
GABARAP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11337, 56486 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95166, Q9DCD6 | |
Gabarap | |
GABARAP Fusion Protein Ag18209 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |