Learn More
Furan-2-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
78.00 € - 312.00 €
Chemical Identifiers
CAS | 374790-93-9 |
---|---|
Molecular Formula | C10H15BO3 |
Molecular Weight (g/mol) | 194.04 |
MDL Number | MFCD02094008 |
InChI Key | GWEWQKZABZXLJH-UHFFFAOYSA-N |
Synonym | 2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol ester, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, furan-2-boronic acid pinacol cyclic ester, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, zlchem 693, pubchem18440 |
PubChem CID | 11356013 |
IUPAC Name | 2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=CO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304985
|
Thermo Scientific Alfa Aesar
L18366.03 |
1 g |
78.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314985
|
Thermo Scientific Alfa Aesar
L18366.06 |
5 g |
312.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
374790-93-9 | |
194.04 | |
GWEWQKZABZXLJH-UHFFFAOYSA-N | |
11356013 | |
CC1(C)OB(OC1(C)C)C1=CC=CO1 |
C10H15BO3 | |
MFCD02094008 | |
2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol ester, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, furan-2-boronic acid pinacol cyclic ester, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, zlchem 693, pubchem18440 | |
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
374790-93-9 | |
1.472 | |
9045028 | |
GWEWQKZABZXLJH-UHFFFAOYSA-N | |
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
11356013 | |
98% |
C10H15BO3 | |
MFCD02094008 | |
2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol ester, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, furan-2-boronic acid pinacol cyclic ester, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, zlchem 693, pubchem18440 | |
CC1(C)OB(OC1(C)C)C1=CC=CO1 | |
194.04 | |
194.04 | |
Furan-2-boronic acid pinacol ester |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.