missing translation for 'onlineSavingsMsg'
Learn More

Fumaryl chloride, 95%, Thermo Scientific Chemicals

52.90 € - 150.00 €

Chemical Identifiers

CAS 627-63-4
Molecular Formula C4H2Cl2O2
Molecular Weight (g/mol) 152.958
MDL Number MFCD00000733
InChI Key ZLYYJUJDFKGVKB-OWOJBTEDSA-N
Synonym fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
PubChem CID 5325504
IUPAC Name (E)-but-2-enedioyl dichloride
SMILES C(=CC(=O)Cl)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491908
View Documents
Thermo Scientific Alfa Aesar
A14540.14
25 g
52.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
11401918
View Documents
Thermo Scientific Alfa Aesar
A14540.22
100 g
150.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

Solubility
It reacts with water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, active metals and bases.
Specifications

Chemical Identifiers

627-63-4
152.958
ZLYYJUJDFKGVKB-OWOJBTEDSA-N
5325504
C(=CC(=O)Cl)C(=O)Cl
C4H2Cl2O2
MFCD00000733
fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
(E)-but-2-enedioyl dichloride

Specifications

627-63-4
1.413
73°C (163°F)
C4H2Cl2O2
MFCD00000733
1721342
fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
ZLYYJUJDFKGVKB-OWOJBTEDSA-N
(E)-but-2-enedioyl dichloride
5325504
95%
-2°C
159°C to 161°C
Acrid
1.5
UN1780
Moisture sensitive
It reacts with water.
C(=CC(=O)Cl)C(=O)Cl
152.958
152.97
Fumaryl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312+H332-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FUMARYL CHLORIDE

missing translation for 'einecsNumber' : 211-005-4

missing translation for 'rtecsNumber' : LT2800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only