missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluorotetrachloroethane, SPEX CertiPrep™

Product Code. 16360038
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Difluorotetrachloroethane76-12-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 76-12-0, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C2Cl4F2
InChI Key UGCSPKPEHQEOSR-UHFFFAOYSA-N
SMILES C(C(F)(Cl)Cl)(F)(Cl)Cl
IUPAC Name 1,1,2,2-tetrachloro-1,2-difluoroethane
Molecular Weight (g/mol) 203.819
PubChem CID 6427
Solvent or Matrix Methanol
Chemical Name or Material 1,2-Difluorotetrachloroethane
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'