missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18592-1-AP-150UL
This item is not returnable.
View return policy
Description
FOXO1 (FKHR, ForKHead Box 01) is a member in a subfamily of the forkhead homeotic gene family of transcription factors. Recent experiments have shown that FOXO1 can act as either a coactivator or a corepressor of nuclear receptor activity that is mediated through the LXXLL motif found in the carboxyl terminal region of the FKHR protein. Association of FOXO1 with PAX3 has been implicated in alveolar rhabdomyosarcoma. Recent studies link the anti-tumor activity of FOXO1, the process of autophagy and myogenic growth and differentiation. FOXO1 is the main target of insulin signaling that regulated the metabolic homeostasis in response to oxidative stress. FOXO1 binds to the insulin response element (IRE) with consensus sequence 5′-TT[G/A]TTTTG-3′ and the related Daf-16 family binding element (DBE) with consensus sequence 5′- TT[G/A]TTTAC-3′. FOXO1 is a regulator of redox balance, osteoblast numbers and controls bone mass. Further, FOXO1 orchestrates the endocrine function of the skeleton in regulating glucose metabolism, and suppresses the transcriptional activity of RUNX2, an upstream activator of osteocalcin/BGLAP. In hepatocytes, FOXO1 promotes gluconeogenesis by acting together with PPARGC1A to activate the expression of genes such as IGFBP1, G6PC and PPCK1. FOXO1 is an important regulator of cell death acting downstream of CDK1, PKB/AKT1 and SKT4/MST1.Specifications
FOXO1 | |
Polyclonal | |
Unconjugated | |
FOXO1 | |
FKH1, FKHR, forkhead box O1, Forkhead box protein O1, Forkhead box protein O1A, Forkhead in rhabdomyosarcoma, FOXO1, FOXO1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2308, 56458, 84482 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G3V7R4, Q12778, Q9R1E0 | |
Foxo1 | |
FOXO1 Fusion Protein Ag13296 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |