Learn More
Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers
For dispensing of all chemicals - including solvents - that do not attack polypropylene and don't crystallize. Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers come with a glass piston.
Specifications
Accuracy 0.60%
Color Natural
View More Specs
Description
Description
For dispensing of all chemicals - including solvents - that do not attack polypropylene and don't crystallize. Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers come with a glass piston.
  • Visible dispensing procedure
  • Easy to use volume setting system
  • Valve block made of polypropylene with FIX adapter
Specifications
Specifications
0.60%
Natural