missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
35.64 € - 292.00 €
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10391731
|
Thermo Scientific Acros
410775000 |
500 mL | Plastic bottle |
35.64 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697021
|
Thermo Scientific Acros
410770025 |
2.5 L | Plastic bottle |
108.70 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904891
|
Thermo Scientific Acros
410770100 |
10 L | Plastic drum |
292.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-18-6 | |
46.025 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
Specifications
64-18-6 | |
101.0°C | |
CH2O2 | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
88% | |
1.2200g/mL | |
Authentic | |
1.3711 to 1.3731 | |
1.22 |
8.0°C | |
88% | |
HCO2H | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
ACS Reagent | |
60°C | |
Liquid | |
0.002% max. | |
Formic acid |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Formic acid, 88%
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals