Learn More
Formic acid-d2, for NMR, +99% atom D, 95% solution in D2O, Thermo Scientific Chemicals
66.43 € - 397.98 €
Chemical Identifiers
CAS | 920-42-3 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 48.037 |
MDL Number | MFCD00037363 |
InChI Key | BDAGIHXWWSANSR-PFUFQJKNSA-N |
Synonym | formic acid-d2, 2h formic 2 acid, dcood, 2h2 formic acid, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide, formic acid-d2, 95 wt. % in d2o, 98 atom % d |
PubChem CID | 123092 |
IUPAC Name | deuterio deuterioformate |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10742194
|
Thermo Scientific Acros
233400010 |
1 g | Glass Bottle |
66.43 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033140
|
Thermo Scientific Acros
233400100 |
10 g | Glass Bottle |
397.98 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
920-42-3 | |
48.037 | |
BDAGIHXWWSANSR-PFUFQJKNSA-N | |
123092 | |
C(=O)O |
CH2O2 | |
MFCD00037363 | |
formic acid-d2, 2h formic 2 acid, dcood, 2h2 formic acid, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide, formic acid-d2, 95 wt. % in d2o, 98 atom % d | |
deuterio deuterioformate |
Specifications
Liquid | |
1.2700g/mL | |
50°C | |
920-42-3 | |
101.0°C | |
DCOOD | |
formic acid-d2, 2h formic 2 acid, dcood, 2h2 formic acid, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide, formic acid-d2, 95 wt. % in d2o, 98 atom % d | |
BDAGIHXWWSANSR-PFUFQJKNSA-N | |
deuterio deuterioformate | |
123092 | |
≥94.0% | |
Formic acid-d2, 99+ atom % D (95% in D2O) |
Colorless | |
99.0% min. | |
1.27 | |
5.9°C | |
CH2O2 | |
MFCD00037363 | |
Solubility in water: miscible. | |
C(=O)O | |
48.037 | |
48.04 | |
NMR |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTRE or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-057-3
- Formic acid-d2
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.