missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, ≥99.0% (GC), Reagent Grade, Honeywell™
Reagent Grade,≥99.0% (GC)
20.83 € - 287.90 €
Chemical Identifiers
CAS | 75-12-7 |
---|---|
Molecular Formula | CH3NO |
Molecular Weight (g/mol) | 45.04 |
MDL Number | MFCD00007941 |
InChI Key | ZHNUHDYFZUAESO-UHFFFAOYSA-N |
Synonym | methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 |
PubChem CID | 713 |
ChEBI | CHEBI:48431 |
IUPAC Name | formamide |
SMILES | NC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15648490
|
Honeywell Research Chemicals
F7503-5ML |
5 mL | Glass Bottle | ||||||
15697870
|
Honeywell Research Chemicals
F7503-100ML |
100 mL | Glass Bottle | ||||||
15627880
|
Honeywell Research Chemicals
F7503-250ML |
250 mL | Glass Bottle | ||||||
15637880
|
Honeywell Research Chemicals
F7503-500ML |
500 mL | Glass Bottle | ||||||
15607880
|
Honeywell Research Chemicals
F7503-1L |
1 L | Glass Bottle | ||||||
15617880
|
Honeywell Research Chemicals
F7503-2.5L |
2.5 L | Poly Bottle | ||||||
15638490
|
Honeywell Research Chemicals
F7503-4L |
4 L | Poly Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
75-12-7 | |
45.04 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
713 | |
formamide |
CH3NO | |
MFCD00007941 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
CHEBI:48431 | |
NC=O |
Specifications
4.0 to 5.0 (at 20°C) | |
75-12-7 | |
1.134g/mL at 25°C | |
175°C (347°F) | |
Weak | |
CH3NO | |
MFCD00007941 | |
NONH for all modes of transport | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
formamide | |
713 | |
45.04g/mol | |
Formamide |
1°C to 3°C | |
Colorless | |
210°C (1013 hPa) | |
932°F | |
Liquid | |
HCONH2 | |
0.08mmHg (20°C), 30mmHg (129°C) | |
505995 | |
Fully miscible in water | |
NC=O | |
45.04 | |
CHEBI:48431 | |
≥99.0% (GC) |
Safety and Handling
P201-P260-P280-P308 + P313
missing translation for 'rtecsNumber' : 200-842-0
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Formamide
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.