Learn More
Thermo Scientific™ Fluoro-Max™ Fluorescent Carboxylate-Modified Particles
Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence.
Brand: Thermo Scientific™ 93470520010150
Description
Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence with Thermo Scientific™ Fluoro-Max Fluorescent Carboxylate-Modified Particles. Made by dyeing Thermo Scientific™ OptiLink™ carboxylate-modified particles with europium chelate, particles have been specifically designed for membrane or automated fluorometric-based applications. With an extremely broad Stokes shift, Fluoro-Max europium chelate particles help prevent nonspecific fluorescence interference.
Fluoro-Max Fluorescent Particles are made from carboxylate-modified polystyrene particles containing encapsulated europium chelate dye. They are available in standard 0.1, 0.2, or 0.3μm diameters. When these particles excite at 333nm and emit at 613nm, they do so with a very long lifetime of approximately 0.5 milliseconds. In fact, this is about 10,000 to 100,000 times the lifetime of most fluorophores. The extended lifetime of europium chelate particles allows them to be used in a wide range of time resolved fluorescence applications. In addition, they can be counted on to lower the detection limits of fluorescent assays severalfold as they eliminate background interference from relatively short-lived matrix fluorescence.- Internally dyed to ensure maximum color brilliance and color saturation, resulting in greater sensitivity and enhanced readability of the assay. Eliminates problems associated with enzyme-based assays
- Encapsulated dye prevents leaching in aqueous media, and provides optimum surface immunoreactivity
- Dyeing process causes little or no inhibition of adsorption or covalent couping of immunoglobulin G, resulting in superior binding of protein to the particles
- Suitable for a variety of applications such as clinical diagnostics, immuno/histological studies and research
Recommended for:
Clinical diagnostics, molecular biology and lateral flow, turbidimetric, and slide aggulation assays

Specifications
0.05% Sodium Azide | |
1% solids | |
Fluorescent red dyed polystyrene microspheres in water | |
Polystyrene or Polystyrene with Coploymer Grafted Surface | |
Europium Chelate | |
20 A²/group | |
5 mL | |
Carboxylate-Modified | |
0.2 μm | |
0.2 μm |
Carboxylate-modified | |
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Shipment at ambient temperature. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. | |
White | |
Fluroescent Carboxylate-Modified Particle | |
Package Insert Sheet containing Certicate of Analysis | |
1.06g/cm³ | |
Product stable for a minimum of 36 months | |
<5% CV | |
0.2 μm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
256456 | Certificate of Analysis | 29/01/2025 | 93470520010150 |
252465 | Certificate of Analysis | 29/01/2025 | 93470520010150 |
253282 | Certificate of Analysis | 29/01/2025 | 93470520010150 |
259884 | Certificate of Analysis | 29/01/2025 | 93470520010150 |
260213 | Certificate of Analysis | 29/01/2025 | 93470520010150 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.