missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein diacetate, 97%, pure

51.56 € - 598.98 €

Chemical Identifiers

CAS 596-09-8
Molecular Formula C24H16O7
Molecular Weight (g/mol) 416.39
MDL Number MFCD00005062
InChI Key CHADEQDQBURGHL-UHFFFAOYSA-N
Synonym fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetate
PubChem CID 65047
IUPAC Name (6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10134450
SDS View Documents
Thermo Scientific Chemicals
191660050
5 g Glass bottle
51.56 €
5g
Please to purchase this item. Need a web account? Register with us today!
10067432
SDS View Documents
Thermo Scientific Chemicals
191660250
25 g Glass bottle
173.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
10189552
SDS View Documents
Thermo Scientific Chemicals
191661000
100 g Glass bottle
598.98 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

596-09-8
416.39
CHADEQDQBURGHL-UHFFFAOYSA-N
65047
CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3
C24H16O7
MFCD00005062
fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetate
(6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate

Specifications

596-09-8
Yellow
96% min. (HPLC)
MFCD00005062
fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetate
CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3
416.39
416.39
Pure
Fluorescein diacetate
200.0°C to 205.0°C
Authentic
C24H16O7
19,227
CHADEQDQBURGHL-UHFFFAOYSA-N
(6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
65047
97%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 209-877-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.