missing translation for 'onlineSavingsMsg'
Learn More

IKA™ R 472 Floor stand

Use this mobile floor stand for a variety of laboratory applications. IKA Floor Stands are specially designed for the overhead stirrer RW 47 digital and can be adapted for use with other instruments.

Brand:  IKA™ 0000738700

Product Code. 10202411

  • 5645.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dimensions (D x W): 80 x 80mm 
  • Height: 2.020mm 
  • Stroke 980 to 1.860mm

Order Info

HeightMetric

Specifications

Specifications

2020 mm
1 Ea.
Floor Stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.