Learn More
Ansell Flexitril™ L27 Nitrile-Coated Chemical/Liquid Protection Gloves
Product Code. 16855340
Click to view available options
Size:
7
8
9
10
Gloves are flexible and comfortable while also providing protection against harmful chemicals, oils, and greases.
  • Gloves combine flexibility, dexterity, and mechanical resistance
  • Coating is resistant to chemicals, oils, greases, gasoline, petrol, alcohol, alkali, and salts
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Green
Material Nitrile
Size 8
EN388 Standard 3121B
Glove Cuff Style Gauntlet
Liner Material Interlock Cotton
Lining Lined
Resistant To Chemicals, Oil, Grease, Gasoline, Petrol, Alcohols, Alkalis, and Salts
Series L27
Length (Metric) 270 mm