missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG
This Antibody Labeling Kit conjugates CoraLite™ 488 into Rabbit IgG antibodies.
90.00 € - 889.00 €
Specifications
Content And Storage | 1 Year at -20°C or 6 Months at +4°C, Avoid Light |
---|---|
Excitation/Emission | 500 nm/520 nm |
Labeling Target | IgG |
Species | Rabbit |
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
17741509
|
Proteintech™
KFA001-10 |
10 μL CoraLite™ 488 FlexLinker for Rabbit IgG, 20 μL FlexQuencher for Rabbit IgG, 100 μL FlexBuffer, Manual, Protocol Magnet |
90.00 €
10 reactions |
In Stock
Log in to see stock. |
|||||
17761509
|
Proteintech™
KFA001-50 |
50 μL CoraLite™ 488 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet |
419.00 €
50 reactions |
In Stock
Log in to see stock. |
|||||
17751509
|
Proteintech™
KFA001-200 |
4 x 50 μL CoraLite™ 488 FlexLinker for Rabbit IgG, 4 x 100 μL FlexQuencher for Rabbit IgG, 4 x 500 μLFlexBuffer, Manual, Protocol Magnet |
889.00 €
200 reactions |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to rabbit IgG primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
500 nm/520 nm | |
IgG | |
Rabbit |