missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Two-Hole Rubber Stoppers

Assorted sizes 00 to 15

48.50 €

Specifications

Color Black
Material Natural Rubber
Product Type Tapered Stopper (2 holes)
For Use With (Application) General purpose use
Total Positions 2
View More Specs

Products 1
Product Code Brand Length (Metric) Size Stopper Number Diameter (Metric) Top Price Quantity & Availability  
Product Code Brand Length (Metric) Size Stopper Number Diameter (Metric) Top Price Quantity & Availability  
11756078
View Documents
Fisherbrand™
62991-73
2.5 cm 2 2 2 cm N/A N/A N/A
Description

Description

Assorted sizes 00 to 15

  • Hole size: 3 to 5mm

The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).

Specifications

Specifications

Black
Tapered Stopper (2 holes)
2
Natural Rubber
General purpose use
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Two-Hole Rubber Stoppers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.