missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety-Labeled Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
1000 mL
For Use With (Application):
Acetone
Deionized Water
Distilled Water
Ethanol
Isopropanol
Toluene
Description
Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.
- Easy-to-read printed bottles
- Wide mouth 53 mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 mm, Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Use only with solvents compatible with LDPE
Order Info
Available in cases of six, all with the same label, or in a six-bottle assortment: one 500mL bottle each of acetone, ethanol, isopropanol, methanol, toluene and water.
Specifications
Specifications
Capacity | 17 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 17 oz. |
Color | Natural |
For Use With (Application) | Ethanol |
Material | Polyethylene |
Sterility | Non-sterile |
Closure Color | Natural |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Closure Included | Yes |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Safety-Labeled Wash Bottles >
Spot an opportunity for improvement?Share a Content Correction