missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids

Fit standard spectrophotometer cell holders

Brand:  Fisherbrand™ 14958107

 View more versions of this product

Product Code. 15296003


Includes: PTFE lid or stopper
This item is not returnable. View return policy

Description

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Micro Fluorescence Cell
12 μL
PTFE
Ea., 2 Ea./Cs.
200 to 2500 nm
Standard spectrophotometer cell holder
Transmission from far ultraviolet through the infrared
Center H: 15mm
Quartz
1.5mm
1.5 mm
45 mm
12.5 mm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids > w/PTFE lid; Micro; 12μL; Center H: 15mm; Path L: 1.5mm; Internal W: 1.5mm; Ea.; 2 Ea./Cs.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.