Learn More
Bel-Art™ SP Scienceware™ All-Metal Quick-Disconnects
Inline hose connector with automatic shutoff on disconnect to prevent leakage
Brand:  Bel-Art™ F19723-0802
 View more versions of this product
Product Code. 10279681
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Chrome-plated brass hookups Compatible with most tubing
  • Operates at temperatures to 82°C (180°F), pressures to 250psi (1.7MPa) and vacuum to 30 in.Hg
Order Info
Order male and female connectors separately so tubing of the same or different size can be connected.
Specifications
Specifications
Quick Disconnect, Female Coupling
3.2 mm
3.2mm
0.125 in.
Metal
250 psi