Learn More
Fisherbrand™ Preparative/Marker Comb for Owl™ P2-CST Multiple Gradient Gel Caster
Preparative/Marker Comb for Owl™ P2-CST Multiple Gradient Gel Caster
Brand: Fisherbrand™ P2-PREP
This item is not returnable.
View return policy
Description
- For use with Owl P2-CST Multiple Gradient Gel Caster
Specifications
Owl P2-CST Multiple Gradient Gel Caster | |
10 x 20 cm | |
0.15 cm | |
7.8 in. | |
7.8 in. | |
20 cm |
Preparative/Marker Comb | |
2 | |
448/155 μL | |
20 cm | |
5.8/0.18 in. | |
14.81/0.47 cm |
Product Content Correction
Product Title
Fisherbrand™ Preparative/Marker Comb for Owl™ P2-CST Multiple Gradient Gel Caster > Preparative/Marker comb; For gel size: 20L x 20cmW
Spot an opportunity for improvement?Share a Content Correction