missing translation for 'onlineSavingsMsg'
Learn More

Freezer ultra-low Isotemp -86°C upright 650L

Brand:  Fisherbrand

Freezer ultra-low Isotemp -86°C upright 650L

Code : 46

Additional Details : Weight : 0.01000kg

Product Code. 12609275

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers