Learn More
Assorted pH Buffer Pouches, Fisherbrand™
Economical, accurate, convenient
Brand: Fisherbrand 35653-04
This item is not returnable.
View return policy
Description
Economical, accurate, convenient
- ±0.01pH accuracy at 25°C (77°F)
- Pack of 20 pouches: five pouches each of pH 4, 7, and 10 buffers and rinse solution
Certifications
All solutions are certified traceable to NIST™ standards.
Specifications
4; 10; 7 | |
20 pouches | |
Buffer |
Product Content Correction