missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ OptiGrow™ LB Broth Miller Microbiological Media
Classic media in ready-to-use liquid form
Brand: Fisher BioReagents™ BP1430-10B1
This item is not returnable.
View return policy
Description
- Sterilized by autoclaving
- For high yield of bacterial culture
- No dusty powders to cause environmental concerns
- Fully automatic manufacturing process eliminates unexpected contamination
- LB Broth. Standard bacteriological nutrient medium for the propagation of E. coli for purposes of strain maintenance (sub-culturing), cloning, plasmid propagation, and protein expression
- Super Broth. Specifically developed to increase the yield of high copy number plasmids derived from suspension cultures of E. coli strains
- Terrific Broth. Supports higher cell densities than LB medium, increases plasmid DNA yield
Order Info
Supplied in case of ten 1L bottles
Specifications
Yellow | |
United States | |
10 x 1000 mL | |
10 x 1 L |
Liquid | |
LB Broth, Miller | |
Bottle |
Safety and Handling
Product Identifier
- OptiGrow LB Broth Miller Liquid (Luria-Bertani Broth)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction