missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Bioreagents Taq DNA Polymerase 250U (5U/µL) Fisher BioReagents
Brand: Fisher Bioreagents FB6111
Taq DNA Polymerase 250U (5U/µL) Fisher BioReagents
Code : 32
Additional Details : Weight : 0.06800kg