Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Perform PCR and other purity demanding applications with these biocon pipette tips, certified free from endotoxin, DNA, DNase and RNase.
Brand: Thermo Scientific™ 94053200
This item is not returnable.
View return policy
Description
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:- Individually packed
- Three models in volume range 0.2-1000 μL
- Presterilized with gamma radiation
- Includes certificate of quality
Recommended for: PCR applications, molecular biology
Specifications
Finntip BioCon 250, Non-Filtered, Sterile, | |
Non-filtered | |
Finntip BioCon 250 | |
Thermo Scientific FinnTip BioCon 250 |
250 μL | |
Sterile | |
ISO 9001, ISO 14001, ISO 13485 | |
FinnTip Biocon |
Product Content Correction
Product Title
Thermo Scientific™ Finntip™ Biocon Pipette Tips > 0.5 to 250 μL
Spot an opportunity for improvement?Share a Content Correction