Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
281.00 € - 915.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15332167
|
Thermo Scientific™
010-1072 |
30mL Oak Ridge Tube |
915.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15342167
|
Thermo Scientific™
010-1074 |
16mL Oak Ridge Tube |
900.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15362167
|
Thermo Scientific™
010-1117 |
10mL Vacutainer Tube |
705.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15322167
|
Thermo Scientific™
010-1119 |
100mL Oak Ridge Tube |
705.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15372167
|
Thermo Scientific™
010-1138 |
3mL Vacutainer Tube |
900.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15209653
|
Thermo Scientific™
010-1410 |
15 mL Conical Tube |
281.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.