missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VACUUBRAND™ FFKM O-ring
69.85 € - 296.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11909741
|
VACUUBRAND™
20638741 |
14 x 4mm |
97.65 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
11989731
|
VACUUBRAND™
20638740 |
28 x 5mm |
296.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
11919741
|
VACUUBRAND™
20638742 |
8 x 3mm |
69.85 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
Specifications
O-Ring | |
FFKM | |
1Pack |